Läs mer
ent-Montelukast Sodium Salt, TRC
Produktkod. 30011755
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011755 10 mg
30006524 50 mg
30009284 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011755 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM56799510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 190078-45-6
Molekylformel C35 H35 Cl N O3 S . Na
Molekylvikt (g/mol) 608.17
Synonym Cyclopropaneacetic acid, 1-[[[(1S)-1-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, sodium salt (1:1), Cyclopropaneacetic acid, 1-[[[(1S)-1-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, monosodium salt (9CI), Cyclopropaneacetic acid, 1-[[[1-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, monosodium salt, [S-(E)]-, L 768232, (S)-Montelukast, ent-Montekulast Sodium Salt, Montelukast Sodium Imp. A (EP) as Sodium Salt
IUPAC-namn sodium;2-[1-[[(1S)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate
LEDER [Na+].CC(C)(O)c1ccccc1CC[C@H](SCC2(CC(=O)[O-])CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3
Specifikationer
Kemiskt namn eller material ent-Montelukast sodium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C35H36ClNO3S.Na/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29;/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39);/q;+1/p-1/b15-10+;/t32-;/m0./s1
Formel vikt 607.1924
Procent renhet >95%