Läs mer
ent NAP 226-90, TRC
Produktkod. 30179736
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179736 100 mg
30179751 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179736 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN325005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 851086-95-8
Molekylformel C10 H15 N O
Molekylvikt (g/mol) 165.23
Synonym Phenol, 3-[(1R)-1-(dimethylamino)ethyl]-, 3-[(1R)-1-(Dimethylamino)ethyl]phenol, (R)-3-[1-(Dimethylamino)ethyl]phenol
IUPAC-namn 3-[(1R)-1-(dimethylamino)ethyl]phenol
LEDER C[C@@H](N(C)C)c1cccc(O)c1
Specifikationer
Kemiskt namn eller material ent NAP 226-90
Kvantitet 100 mg
InChI formel InChI=1S/C10H15NO/c1-8(11(2)3)9-5-4-6-10(12)7-9/h4-8,12H,1-3H3/t8-/m1/s1
Formel vikt 165.1154