Läs mer
Entecavir-13C2,15N, TRC
Produktkod. 30012421
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30012421 0.5 mg 0.5mg
30008945 5 mg -
30005045 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012421 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5589020.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329796-53-3
Molekylformel C1013C2H15N415NO3
Molekylvikt (g/mol) 280.26
Synonym 2-Amino-9-[4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one-13C2,15N, Baraclude-13C2,15N, BMS 200475-13C2,15N, SQ 34676-13C2,15N;
LEDER O=[13C]1C([15N]=CN2[C@H]3C[C@H](O)[C@@H](CO)C3=C)=[13C]2N=C(N)N1
Specifikationer
Kemiskt namn eller material Entecavir-13C2,15N
Anteckningar om renhetsgrad HPLC
Kvantitet 0.5 mg
InChI formel InChI=1S/C12H15N5O3/c1-5-6(3-18)8(19)2-7(5)17-4-14-9-10(17)15-12(13)16-11(9)20/h4,6-8,18-19H,1-3H2,(H3,13,15,16,20)
Procent renhet >95%