Learn More
Sartorius Entris™ II Advanced Precision Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA10201I-1CFR
Additional Details : Weight : 4.60000kg
 View more versions of this product
Product Code. 16616243
  • 37480.00 SEK / Each
Estimated Shipment: 05-09-2022
to see stock.
Description
Description
  • Entris™ II Advanced Balances - Quality, Value, and Consistency
  • For accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
Specifications
Specifications
CE Certificate
Verified (France Country-Specific)
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤0.9 seconds
0.2 A
D-sub connector, 9-pin; USB UTL; USB Device
10,200 g
100 mg
Electronic Precision Balance
100/240 V (± 10%)
50/60 Hz