Learn More
EOMES Monoclonal Antibody (WD1928), APC-eFluor™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1766.00 SEK - 4200.00 SEK
Specifications
Antigen | EOMES |
---|---|
Clone | WD1928 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15559586
|
Affymetrix eBioscience
47-4877-41 |
25 Tests |
1766.00 SEK
25 tests |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15569586
|
Affymetrix eBioscience
47-4877-42 |
100 Tests |
4200.00 SEK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This WD1928 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Applications Reported: This WD1928 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This WD1928 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Refer to Best Protocols for Staining Protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. APC-eFluor™ 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.
EOMES Monoclonal antibody specifically detects EOMES in Human, Porcine samples. It is validated for Flow Cytometry.Specifications
EOMES | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
Eomes | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Eomes |
WD1928 | |
Flow Cytometry | |
APC-eFluor 780 | |
Mouse | |
Human, Porcine | |
O95936 | |
100622125, 8320 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.