missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Eosin-5-Isothiocyanate

Product Code. 11554736
Change view
Click to view available options
Kvantitet:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
11554736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11554736 Supplier Molecular Probes™ Supplier No. E18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

eosin-5-isothiocyanate

Bioconjugates made with the amine-reactive eosin-5-isothiocyanate can be used as phosphorescent probes or as photosensitizers. With its high quantum yield (∼0.57) for singlet oxygen generation, eosin and its conjugates can be used as effective photooxidizers of diaminobenzidine (DAB) in high-resolution electron microscopy studies and in correlated fluorescence and electron microscopy applications.
TRUSTED_SUSTAINABILITY

Specifications

Beskrivning Eosin–5-Isothiocyanate, 100mg, Orange, Solid, C21H7Br4NO5S Molecular Formula, 704.97 Molecular Weight, Eosin Label, Amine Chemical Reactivity, 521/544nm Excitation/Emission Wavelength, Used for Labeling Chemistry, Protein Labeling and Crosslinking
Formulering Solid
För användning med (applikation) Labeling chemistry, protein labeling and crosslinking
Kvantitet 100 mg
Produkttyp Eosin-5-Isothiocyanate
Molekylvikt (g/mol) 704.97kDa
Renhet HPLC, 95% at 254nm
Innehåll och lagring -5°C to -30°C
Etikett eller färgämne Eosin
Färg Orange
Emission 544
Excitation 521
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.