Läs mer
Eosine-5-isothiocyanate, TRC
Produktkod. 30373503
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30373503 250 mg
1 options
Produktkod. 30373503 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE561010250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Eosine-5-isothiocyanate
CAS 60520-47-0
Molekylformel C21 H7 Br4 N O5 S
Kvantitet 250 mg
Synonym 2′,4′,5′,7′-Tetrabromo-3′,6′-dihydroxy-5-isothiocyanatospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one (ACI), 5-Isothiocyanato-2-(2,4,5,7-tetrabromo-3-hydroxy-6-oxoxanthen-9-yl)benzoic acid, Eosin 5-isothiocyanate, 2′,4′,5′,7′-Tetrabromo-3′,6′-dihydroxy-5-isothiocyanatospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one (ACI), 5-Isothiocyanato-2-(2,4,5,7-tetrabromo-3-hydroxy-6-oxoxanthen-9-yl)benzoic acid, Eosin 5-isothiocyanate 2′,4′,5′,7′-Tetrabromo-3′,6′-dihydroxy-5-isothiocyanatospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one (ACI), 5-Isothiocyanato-2-(2,4,5,7-tetrabromo-3-hydroxy-6-oxoxanthen-9-yl)benzoic acid, Eosin 5-isothiocyanate, Eosin isothiocyanate
InChI formel InChI=1S/C21H7Br4NO5S/c22-12-4-10-18(14(24)16(12)27)30-19-11(5-13(23)17(28)15(19)25)21(10)9-2-1-7(26-6-32)3-8(9)20(29)31-21/h1-5,27-28H
LEDER O=C1OC2(C3=C(OC4=C2C=C(Br)C(O)=C4Br)C(Br)=C(O)C(Br)=C3)C5=C1C=C(N=C=S)C=C5
IUPAC-namn 2',4',5',7'-tetrabromo-3',6'-dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
Molekylvikt (g/mol) 704.96
Formel vikt 700.68