missing translation for 'onlineSavingsMsg'
Learn More

EPB41L4A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26596-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16875015

  • 4721.67 SEK / 150µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the band 4.1 protein superfamily, including EPB41L4A, are thought to regulate the interaction between the cytoskeleton and plasma membrane (Ishiguro et al., 2000 ).
Specifications

Specifications

EPB41L4A
Polyclonal
Unconjugated
EPB41L4A
Band 4.1 like protein 4A, EPB41L4, EPB41L4A, NBL4, Protein NBL4
Rabbit
Antigen Affinity Chromatography
RUO
13824, 64097
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P52963, Q9HCS5
EPB41L4A
EPB41L4A Fusion Protein Ag23923
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers