Learn More
EPB41L5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558009
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DLLEATIGDV IGASDTMETS QALNDVNVAT RLPGLGEPEV EYETLKDTSE KLKQLEMENS PLLSPRSNID VNINSQEEVV KLTEKC Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 60%.
EPB41L5 Polyclonal antibody specifically detects EPB41L5 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
EPB41L5 | |
Polyclonal | |
Unconjugated | |
EPB41L5 | |
Band 4.1-like protein 5; BE37; EPB41L5; erythrocyte membrane protein band 4.1 like 5; KIAA1548; YMO1; YRT; yurt homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57669 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCM4 | |
EPB41L5 | |
Recombinant protein corresponding to Human EPB41L5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only