missing translation for 'onlineSavingsMsg'
Learn More

EPHA1 Mouse anti-Human, Clone: 1B7C3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60155-2-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877125

  • 4721.67 SEK / 150µL
Estimated Shipment: 20-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene is expressed in some human cancer cell lines and has been implicated in carcinogenesis.
Specifications

Specifications

EPHA1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P21709
EPHA1
Peptide
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1B7C3
Unconjugated
EPHA1
EPH, EPH receptor A1, EPHA1, Ephrin type A receptor 1, EPHT, EPHT1
Mouse
Caprylic Acid Precipitation
RUO
2041
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers