missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ EphA1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547837
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) EphA3, rmEphA8, rmEphA4, recombinant rat (rr) EphA5, rmEphA2, rmEphA6, and rmEphA7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
EphA1 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA1 is expressed in some human cancer cell lines and has been implicated in carcinogenesis.
Specifikationer
| EphA1 | |
| Polyclonal | |
| Unconjugated | |
| EPHA1 | |
| 5730453L17Rik; AL033318; Embryonic stem cell kinase; EPH; EPH receptor A1; EPH tyrosine kinase; EPH tyrosine kinase 1; EPHA1; ephrin receptor EphA1; ephrin type-A receptor 1; EPHT; EPHT1; erythropoietin-producing hepatoma amplified sequence; erythropoietin-producing hepatoma receptor; Esk; hEpha1; mEpha1; MGC163163; oncogene EPH; soluble EPHA1 variant 1; soluble EPHA1 variant 2; tyrosine-protein kinase receptor EPH; Tyrosine-protein kinase receptor ESK | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 2041 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P21709 | |
| EPHA1 | |
| Mouse myeloma cell line NS0-derived recombinant human EphA1 Lys26-Glu547. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering