missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EPHA7 (Human) Recombinant Protein
Human EPHA7 full-length ORF ( AAH27940.1, 1 a.a. - 279 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002045-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MVFQTRYPSWIILCYIWLLRFAHTGEAQAAKEVLLLDSKAQQTELEWISSPPNGWEEISGLDENYTPIRTYQVCQVMEPNQNNWLRTNWISKGNAQRIFVELKFTLRDCNSLPGVLGTCKETFNLYYYETDYDTGRNVRENLYVKIDTIAADESFTQGDLGERKMKLNTEVREIGPLSKKGFYLAFQDVGACIALVSVKVYYKKCWSIIENLAIFPDTVTGSEFSSLVEVRGTCVSSAEEEAENAPRMHCSAEGEWLVPIGKCICKAGYQQKGDTCECK
Specifications
AAH27940.1 | |
EPH receptor A7 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EPHA7 | |
GST |
2045 | |
Wheat germ expression system | |
10 μg | |
EHK3, HEK11 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |