missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHB4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20883-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene binds to ephrin-B2 and plays an essential role in vascular development.Specifications
EPHB4 | |
Polyclonal | |
Unconjugated | |
EPHB4 | |
EPH receptor B4, EPHB4, Ephrin type B receptor 4, HTK, MYK1, TYRO11, Tyrosine protein kinase TYRO11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13846, 2050, 686310 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54760, P54761 | |
Ephb4 | |
EPHB4 Fusion Protein Ag10033 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |