missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EPHB4 Recombinant Human Protein

EPHB4 is a receptor tyrosine kinase that belongs to the ephrin receptor family.

Brand:  Invitrogen™ PR4688B

Additional Details : Weight : 0.01000kg

Product Code. 15498413

  • 15570.00 SEK / 100µg
Estimated Shipment: 11-02-2022

Please to purchase this item. Need a web account? Register with us today!


Descripción

Descripción

Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation.

Especificaciones

Especificaciones

NP_004435
Kinase Assay
Frozen
50.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
EPH Receptor Kinase Family
Yes
Baculovirus
Protein Biology, Enzyme & Protein Activity Assays, Kinase Activity, Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
EPHB4, TYRO11
Human
His Tag
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales