Beskrivning
Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Human samples. It is validated for Western Blot
Specifikationer
Specifikationer
| Antigen | Ephrin-A1 |
| Användningsområden | Western Blot |
| Klassificering | Monoclonal |
| Klona | CL6501 |
| Konjugera | PerCP |
| Formulering | PBS |
| Gene Alias | ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 |
| Värd art | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P20827, with the exact immunogen sequence remaining proprietary. |
| Reningsmetod | Protein A purified |
| Visa mer |