missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHX2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10833-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternatively spliced transcript variants have been described.Specifications
EPHX2 | |
Polyclonal | |
Unconjugated | |
EPHX2 | |
CEH, Cytosolic epoxide hydrolase, EPHX2, Epoxide hydratase, Epoxide hydrolase 2, SEH, Soluble epoxide hydrolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2053 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P34913 | |
EPHX2 | |
EPHX2 Fusion Protein Ag1283 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |