Läs mer
Epi-Lacosamide, TRC
Produktkod. 30133408
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133408 25 mg
30138257 250 mg
17939639 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133408 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58879525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 175481-37-5
Molekylformel C13H18N2O3
Molekylvikt (g/mol) 250.29
Synonym Propanamide, 2-(acetylamino)-3-methoxy-N-(phenylmethyl)-, (S)-, (2S)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)propanamide, SPM 6953, (2S)-2-(Acetylamino)-N-benzyl-3-methoxypropanamide, Lacosamide Imp. A (EP), (2S)-2-(Acetamido)-N-benzyl-3-methoxypropanamide, (S)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)propanamide, (2S)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)propanamide, SPM 6953;
IUPAC-namn (2S)-2-acetamido-N-benzyl-3-methoxypropanamide
LEDER COC[C@H](NC(=O)C)C(=O)NCc1ccccc1
Specifikationer
Kemiskt namn eller material Epi-Lacosamide
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C13H18N2O3/c1-10(16)15-12(9-18-2)13(17)14-8-11-6-4-3-5-7-11/h3-7,12H,8-9H2,1-2H3,(H,14,17)(H,15,16)/t12-/m0/s1
Formel vikt 250.1317
Procent renhet >95%