Läs mer
Epiberberine Chloride, TRC
Produktkod. 17943235
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17943235 100 mg
17953535 1 g
17933335 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17943235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE578300100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 889665-86-5
Molekylformel C20 H18 N O4 . Cl
Molekylvikt (g/mol) 371.81
Synonym 7,8,13,13a-Tetradehydro-2,3-dimethoxy-9,10-(methylenedioxy)berbinium Chloride
LEDER [Cl-].COc1cc2CC[n+]3cc4c5OCOc5ccc4cc3c2cc1OC
Specifikationer
Kemiskt namn eller material Epiberberine Chloride
Kvantitet 100 mg
InChI formel InChI=1S/C20H18NO4.ClH/c1-22-18-8-13-5-6-21-10-15-12(3-4-17-20(15)25-11-24-17)7-16(21)14(13)9-19(18)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1
Formel vikt 371.0924