Läs mer
Epiklohydrin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Epichlorohydrin |
| CAS | 106-89-8 |
| Anteckningar om renhetsgrad | GC |
| Molekylformel | C3 H5 Cl O |
| Kvantitet | 500 g |
| Synonym | Oxirane, (chloromethyl)- (9CI), Propane, 1-chloro-2,3-epoxy- (6CI,8CI), 2-(Chloromethyl)oxirane, (Chloromethyl)ethylene oxide, (Chloromethyl)oxirane, (RS)-Epichlorhydrin, (±)-Epichlorohydrin, 1,2-Epoxy-3-chloropropane, 1-Chloro-2,3-epoxypropane, 10: PN: WO2017021334 SEQID: 16 claimed sequence, 2,3-Epoxypropyl chloride, 3-Chloro-1,2-epoxypropane, 3-Chloro-1,2-propylene oxide, 3-Chloropropene-1,2-oxide, 3-Chloropropylene oxide, Chloropropylene oxide, Epicerol, Epichlorohydrin, Epoxychloropropane, Glycerol epichlorohydrin, Glycidyl chloride, J 006, NSC 6747, dl-α-Epichlorohydrin, α-Epichlorohydrin, γ-Chloropropylene oxide |
| InChI formel | InChI=1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2 |
| LEDER | ClCC1CO1 |
| IUPAC-namn | 2-(klorometyl)oxiran |
| Molekylvikt (g/mol) | 92.52 |
| Visa mer |