missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epidermal Growth Factor Receptor/EGFR Ab-10 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.
1275.00 SEK - 7425.60 SEK
Specifications
Antigen | Epidermal Growth Factor Receptor/EGFR Ab-10 |
---|---|
Clone | 111.6 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12661237
|
Epredia
MS-378-B0 |
Concentrated | Biotin | 100 μL |
2205.00 SEK
100µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12671237
|
Epredia
MS-378-B1 |
Concentrated | Biotin | 500 μL |
6535.00 SEK
500µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12621247
|
Epredia
MS-378-P1 |
Concentrated | Unconjugated | 500 μL |
4565.00 SEK
500µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12691237
|
Epredia
MS-378-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12601247
|
Epredia
MS-378-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12611247
|
Epredia
MS-378-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651237
|
Epredia
MS-378-B |
Concentrated | Biotin | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12681237
|
Epredia
MS-378-P |
Concentrated | Unconjugated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12661247
|
Epredia
MS-378-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.Host Species: Mouse
Clone: 111.6
Isotype: IgG1
Species Reactivity: Human. Not mouse and rat.
Epitope: Extracellular domain
Immunogen: Extracellular domain of human recombinant EGFR protein
Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)
Positive Control: A431 cells; Skin, Placenta or Squamous cell CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native only)
- Effect on EGF Binding (Inhibitory)
Note:
USA: RUO; Int'l: RUO
Specifications
Epidermal Growth Factor Receptor/EGFR Ab-10 | |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
Mouse | |
EGFR Family | |
Extracellular domain of human recombinant EGFR protein | |
Primary |
111.6 | |
Monoclonal | |
RUO | |
Human | |
IgG1 |