missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Epigen Monoclonal Antibody (187240)

Beskrivning
Reconstitute at 0.5 mg/mL in sterile PBS.
Epigen is an EGF-related polypeptide growth factor that signals through the ErbB receptor-1. Promotes the growth of epithelial cells. May stimulate the phosphorylation of EGFR and mitogen-activated protein kinases. It is produced in several tissues, including the testis, liver, and heart, as well as in certain tumor cells. Epigen is mitogenic for fibroblasts and epithelial cells. Human Epigen is initially synthesized as a glycosylated 14.7 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a mature soluble sequence.
Specifikationer
Specifikationer
| Antigen | Epigen |
| Användningsområden | Western Blot |
| Klassificering | Monoclonal |
| Klona | 187240 |
| Koncentration | 0.5 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with trehalose and no preservative |
| Gen | EPGN |
| Genaccessionsnr. | Q924X1 |
| Gene Alias | 2310069M11Rik; ALGV3072; EPG; EPGN; epigen; epithelial mitogen; epithelial mitogen homolog; PRO9904; RGD1560084; UNQ3072/PRO9904 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?