Learn More
Thermo Scientific Chemicals (-)-Epinephrine (+)-bitartrate salt, 98+%
Catalog Number p-4229198
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
51-42-3
C13H19NO9
333.29
MFCD00035077
YLXIPWWIOISBDD-NDAAPVSOSA-N
L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate
124080922
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1
51-42-3
C13H19NO9
333.29
MFCD00035077
YLXIPWWIOISBDD-NDAAPVSOSA-N
L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate
124080922
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1
(-)-Epinephrine (+)-bitartrate salt, CAS # 51-42-3, also known as adrenaline, is a monoamine and catecholamine hormone.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• (-)-Epinephrine (+)-bitartrate salt, is a hormone
• It is a monoamine and catecholamine
Applications
• (-)-Epinephrine (+)-bitartrate salt is α-adrenergic and β-adrenergic receptor agonist
• It can be used in adipose region to minimize blood loss
• It is potentially a platelet agonist in preparation of optimal platelet aggregometry 96-well plates
TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 155.0°C
Color White to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,3674
Specific Rotation -20° to -24° (20°C, 589nm) (c=4, 1M HCl)
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Formula Weight 333.29
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material (-)-Epinephrine (+)-bitartrate salt
Show More Show Less
compliance-icons
Product Identifier
  • (-)-Epinephrine (+)-bitartrate salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.