Läs mer
Episterol, TRC
Produktkod. 30005056
Change view
Kvantitet:
0.25 mg
1 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005056 1 mg
30011365 10 mg
30012678 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005056 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5887101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 474-68-0
Molekylformel C28H46O
Molekylvikt (g/mol) 398.66
Synonym 24-Methyl-5α-cholesta-7,24(28)-dien-3β-ol, 24-Methylene-5α-cholest-7-en-3β-ol, 24-Methylenecholesta-7,24(28)-dien-3β-ol, Episterin, Episterol, δ7,24(28)-Ergostadienol
LEDER C=C(C(C)C)CC[C@@H](C)[C@@]1([H])CC[C@@]2([H])C3=CC[C@@]4([H])C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@@]21C
Specifikationer
Kemiskt namn eller material Episterol
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C28H46O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h10,18,20-22,24-26,29H,3,7-9,11-17H2,1-2,4-6H3/t20-,21+,22+,24-,25+,26+,27+,28-/m1/s1
Procent renhet >95%