Läs mer
Epivincamine, TRC
Produktkod. 30374523
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30374523 50 mg
1 options
Produktkod. 30374523 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE94544050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3S,14R,16S)-Vincamine
CAS 6835-99-0
Molekylformel C21 H26 N2 O3
Kvantitet 50 mg
Synonym Eburnamenine-14-carboxylic acid, 14,15-dihydro-14-hydroxy-, methyl ester, (3α,14α,16α)-, 14-Epivincamine (7CI,8CI), 1H-Indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine, eburnamenine-14-carboxylic acid deriv., (-)-cis-Epivincamine, 14-epi-Vincamine, 16-Epivincamine, Epivincamine, cis-Epivincamine, (3S,14R,16S)-Vincamine, (3α,14α,16α)-14,15-Dihydro-14-hydroxyeburnamenine-14-carboxylic acid methyl ester, (12R,13aS,13bS)-13a-Ethyl-2,3,5,6,12,13,13a,13b-octahydro-12-hydroxy-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylic acid methyl ester, Methyl 14-hydroxyvincane-14α-carboxylate
InChI formel InChI=1S/C21H26N2O3/c1-3-20-10-6-11-22-12-9-15-14-7-4-5-8-16(14)23(17(15)18(20)22)21(25,13-20)19(24)26-2/h4-5,7-8,18,25H,3,6,9-13H2,1-2H3/t18-,20+,21-/m1/s1
LEDER CC[C@@]12CCCN3CCc4c([C@H]13)n(c5ccccc45)[C@@](O)(C2)C(=O)OC
Molekylvikt (g/mol) 354.443
Formel vikt 354.194