Läs mer
Eplerenone 7-Carboxylic Acid, TRC
Produktkod. 17970625
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17970625 10 mg
17932195 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17970625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58878510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 209253-82-7
Molekylformel C23H28O6
Molekylvikt (g/mol) 400.46
Synonym Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, γ-lactone, (7α,11α,17α)-, (7α,11α,17α)-9,11-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic acid γ-lactone, 9,11α-Epoxy-7α-carboxy-3-oxo-17α-pregn-4-ene-21,17-carbolactone, 7α-Carboxy-9,11α-epoxy-3-oxo-17α-pregn-4-ene-21,17-carbolactone, (2'R)-9,11α-Epoxy-3,5'-dioxo-4',5'-dihydro-3'H-spiro[androst-4-ene-17,2'-furan]-7α-carboxylic acid, O-Demethyleplerenone, 9,11-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic Acid gamma-Lactone, Eplerenone EP Impurity D
IUPAC-namn (1R,2S,9R,10R,11S,14R,15S,17R)-2,15-dimethyl-5,5'-dioxospiro[18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadec-6-ene-14,2'-oxolane]-9-carboxylic acid
LEDER C[C@]12C[C@H]3O[C@@]34[C@@H]([C@@H](CC5=CC(=O)CC[C@]45C)C(=O)O)[C@@H]1CC[C@@]26CCC(=O)O6
Specifikationer
Kemiskt namn eller material Eplerenone 7-Carboxylic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C23H28O6/c1-20-6-3-13(24)9-12(20)10-14(19(26)27)18-15-4-7-22(8-5-17(25)29-22)21(15,2)11-16-23(18,20)28-16/h9,14-16,18H,3-8,10-11H2,1-2H3,(H,26,27)/t14-,15+,16-,18+,20+,21+,22-,23-/m1/s1
Formel vikt 400.1886