Learn More
Abnova™ EPLIN Recombinant Protein
Human EPLIN full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00051474-P01.25ug
Additional Details : Weight : 0.00010kg
Description
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
- Theoretical MW (kDa): 91.52
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MENCLGESRHEVEKSEISENTDASGKIEKYNVPLNRLKMMFEKGEPTQTKILRAQSRSASGRKISENSYSLDDLEIGPGQLSSSTFDSEKNESRRNLELPRLSETSIKDRMAKYQAAVSKQSSSTNYTNELKASGGEIKIHKMEQKENVPPGPEVCITHQEGEKISANENSLAVRSTPAEDDSRDSQVKSEVQQPVHPKPLSPDSRASSLSESSPPKAMKFQAPARETCVECQKTVYPMERLLANQQVFHISCFRCSYCNNKLSLGTYASLHGRIYCKPHFNQLFKSKGNYDEGFGHRPHKDLWASKNENEEILERPAQLANARETPHSPGVEDAPIAKVGVLAASMEAKASSQQEKEDKPAETKKLRIAWPPPTELGSSGSALEEGIKMSKPKWPPEDEISKPEVPEDVDLDLKKLRRSSSLKERSRPFTVAASFQSTSVKSPKTVSPPIRKGWSMSEQSEESVGGRVAERKQVENAKASKKNGNVGKTTWQNKESKGETGKRSKEGHSLEMENENLVENGADSDEDDNSFLKQQSPQEPKSLNWSSFVDNTFAEEFTTQNQKSQDVELWEGEVVKELSVEEQIKRNRYYDEDEDEE
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH01247 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
91.52 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MENCLGESRHEVEKSEISENTDASGKIEKYNVPLNRLKMMFEKGEPTQTKILRAQSRSASGRKISENSYSLDDLEIGPGQLSSSTFDSEKNESRRNLELPRLSETSIKDRMAKYQAAVSKQSSSTNYTNELKASGGEIKIHKMEQKENVPPGPEVCITHQEGEKISANENSLAVRSTPAEDDSRDSQVKSEVQQPVHPKPLSPDSRASSLSESSPPKAMKFQAPARETCVECQKTVYPMERLLANQQVFHISCFRCSYCNNKLSLGTYASLHGRIYCKPHFNQLFKSKGNYDEGFGHRPHKDLWASKNENEEILERPAQLANARETPHSPGVEDAPIAKVGVLAASMEAKASSQQEKEDKPAETKKLRIAWPPPTELGSSGSALEEGIKMSKPKWPPEDEISKPEVPEDVDLDLKKLRRSSSLKERSRPFTVAASFQSTSVKSPKTVSPPIRKGWSMSEQSEESVGGRVAERKQVENAKASKKNGNVGKTTWQNKESKGETGKRSKEGHSLEMENENLVENGADSDEDDNSFLKQQSPQEPKSLNWSSFVDNTFAEEFTTQNQKSQDVELWEGEVVKELSVEEQIKRNRYYDEDEDEE | |
EPLIN/FLJ38853/MGC131726/SREBP3 | |
LIMA1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
51474 | |
EPLIN (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LIMA1 | |
Human | |
Recombinant | |
Solution |