missing translation for 'onlineSavingsMsg'
Läs mer

EPN-oxon, Dr. Ehrenstorfer

Produktkod. 30326536
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30326536 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326536 Supplier Dr. Ehrenstorfer Supplier No. DREC13180200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material EPN Oxon
CAS 2012-00-2
Analyt- eller komponentnamn EPN Oxon
Frakt skick +20°C
Molekylformel C14 H14 N O5 P
Certifikat och efterlevnad ISO/IEC 17025
Synonym EPN-oxon, Phosphonic acid, phenyl-, ethyl 4-nitrophenyl ester (9CI), Phosphonic acid, phenyl-, ethyl p-nitrophenyl ester (6CI, 7CI, 8CI), EPN oxon, EPNO, Ethyl p-nitrophenyl phenylphosphonate, O-Ethyl O-4-nitrophenyl phenylphosphonate, O-Ethyl O-p-nitrophenyl pheny
InChI formel InChI=1S/C14H14NO5P/c1-2-19-21(18,14-6-4-3-5-7-14)20-13-10-8-12(9-11-13)15(16)17/h3-11H,2H2,1H3
LEDER CCOP(=O)(Oc1ccc(cc1)[N+](=O)[O-])c2ccccc2
IUPAC-namn 1-[ethoxy(phenyl)phosphoryl]oxy-4-nitrobenzene
Molekylvikt (g/mol) 307.238
Kvantitet 10 mg
Formel vikt 307.061 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.