Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Epoxiconazole |
| Förpackning | Glass Bottle |
| CAS | 133855-98-8 |
| Synonym | rac-1-{[(2R,3S)-3-(2-chlorophenyl)-2-(4-fluorophenyl)oxiran-2-yl]methyl}-1H-1,2,4-triazole |
| Industry Type | Environmental Analysis |
| Kvantitet | 100 mg |
| Kvalitet | Analytical Standard |