Läs mer
Epoxiconazole, TRC
Produktkod. 18051130
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18051130 1 g
18063040 100 mg
18022740 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18051130 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5891101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 133855-98-8
Molekylformel C17 H13 Cl F N3 O
Molekylvikt (g/mol) 329.76
Synonym 1H-1,2,4-Triazole, 1-[[(2R,3S)-3-(2-chlorophenyl)-2-(4-fluorophenyl)oxiranyl]methyl]-, rel- (9CI), 1H-1,2,4-Triazole, 1-[[3-(2-chlorophenyl)-2-(4-fluorophenyl)oxiranyl]methyl]-, cis-(±)-, BAS 480F, Epoxiconazole, Opal, Opal (fungicide), Opus, rel-1-[[(2R,3S)-3-(2-Chlorophenyl)-2-(4-fluorophenyl)-2-oxiranyl]methyl]-1H-1,2,4-triazole, 1H-1,2,4-Triazole, 1-[[(2R,3S)-3-(2-chlorophenyl)-2-(4-fluorophenyl)-2-oxiranyl]methyl]-, rel-
IUPAC-namn 1-[[rac-(2R,3S)-3-(2-chlorophenyl)-2-(4-fluorophenyl)oxiran-2-yl]methyl]-1,2,4-triazole
LEDER Fc1ccc(cc1)[C@]2(Cn3cncn3)O[C@H]2c4ccccc4Cl
Specifikationer
Kemiskt namn eller material Epoxiconazole
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C17H13ClFN3O/c18-15-4-2-1-3-14(15)16-17(23-16,9-22-11-20-10-21-22)12-5-7-13(19)8-6-12/h1-8,10-11,16H,9H2/t16-,17-/m0/s1
Formel vikt 329.0731
Procent renhet >95%