Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax Filter Pipet Tips
Tips with liquid blocking technology provide additional protection against over pipetting
Brand:  Eppendorf™ 0030077849
Additional Details : Weight : 1.52000kg
 View more versions of this product
Product Code. 15240785
  • 1793.00 SEK / Pack of 960
Estimated Shipment
08-07-2019
Add to basket
Description
Description
  • Sterile tips made of polyethylene
  • White and violet filter layers in the tip retain aerosols and biomolecules
  • Purple filter layer blocks liquids, binds aerosols and does not allow any liquid to pass through
  • Hydrophobic white layer repels drops, splashes and binds aerosols
  • Free of PCR inhibiting additives
  • Provide protection against contamination of pipet and sample
  • Color coded tip boxes simplify matching pipetes with their tips
PCR and microbiology procedures
Specifications
Specifications
Rack
Filtered
Orange
5.5cm
20 to 300μL
Sterile
PE (Polyethylene)
Videos