Läs mer
Eprosartan, TRC
Produktkod. 17994975
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994975 25 mg
17983925 50 mg
17933995 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE59010025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 133040-01-4
Molekylformel C23 H24 N2 O4 S
Molekylvikt (g/mol) 424.51
Synonym (αE)-α-[[2-Butyl-1-[(4-carboxyphenyl)methyl]-1H-imidazol-5-yl]methylene]-2-thiophenepropanoic Acid, (E)-2-Butyl-1-(p-carboxybenzyl)-α-2-thenylimidazole-5-acrylic Acid, Navixen, SKB 108566, SKF 108566, Teveten;
IUPAC-namn 4-[[2-butyl-5-[(E)-2-carboxy-3-thiophen-2-ylprop-1-enyl]imidazol-1-yl]methyl]benzoic acid
LEDER CCCCc1ncc(\C=C(/Cc2cccs2)\C(=O)O)n1Cc3ccc(cc3)C(=O)O
Specifikationer
Kemiskt namn eller material Eprosartan
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C23H24N2O4S/c1-2-3-6-21-24-14-19(12-18(23(28)29)13-20-5-4-11-30-20)25(21)15-16-7-9-17(10-8-16)22(26)27/h4-5,7-12,14H,2-3,6,13,15H2,1H3,(H,26,27)(H,28,29)/b18-12+
Formel vikt 424.1457
Procent renhet >95%