Learn More
EPSTI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553595
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ARSWAYRDSL KAEENRKLQK MKDEQHQKSE LLELKRQQQE QERAKIHQTE HRRVNNAFLD RLQGKSQPGG LEQSGGCWNM NSGNSW Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 70%.
EPSTI1 Polyclonal antibody specifically detects EPSTI1 in Human samples. It is validated for Immunohistochemistry.Specifications
EPSTI1 | |
Polyclonal | |
Unconjugated | |
EPSTI1 | |
2310046K10Rik; 5033415K03Rik; BRESI1; epithelial stromal interaction 1; epithelial stromal interaction 1 (breast); epithelial-stromal interaction protein 1; EPSTI1; RGD1563207; Unknown (protein for IMAGE:7987136) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
94240 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96J88 | |
EPSTI1 | |
Recombinant protein corresponding to Human EPSTI1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only