Learn More
Eppendorf™ epTIPS™ Pipette Tip Reloads
Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.
Brand:  Eppendorf™ 0030073363
Additional Details : Weight : 0.54900kg
 View more versions of this product
Product Code. 10075361
  • 874.50 SEK / Pack of 960
In stock, ready to ship: 1
Add to basket
Description
Description
Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.
  • Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
  • Racks can be autoclaved
  • Also available in PCR* clean quality, free of human DNA, DNase, RNase and PCR* inhibitors
  • 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
Specifications
Eppendorf™ Pipettor
Non-Filtered
3.4cm
Eppendorf™
Universal Fit
0.1 to 10 μL
Autoclavable
n.a.
Rack