Läs mer
Equilenin, TRC
Produktkod. 17943575
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17943575 1 mg
17924695 10 mg
17951885 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17943575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5924001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 517-09-9
Molekylformel C18 H18 O2
Molekylvikt (g/mol) 266.33
Synonym Equilenin, Estra-1,3,5,7,9-pentaen-17-one, 3-hydroxy-, Equilenin (6CI), 3-Hydroxyestra-1,3,5,7,9-pentaen-17-one, (+)-Equilenin, 3-Hydroxyestra-1,3,5(10),6,8-pentaen-17-one, E 400, Equilenine, NSC 9901, d-Equilenin
IUPAC-namn (13S,14S)-3-hydroxy-13-methyl-12,14,15,16-tetrahydro-11H-cyclopenta[a]phenanthren-17-one
LEDER C[C@]12CCc3c(ccc4cc(O)ccc34)[C@@H]1CCC2=O
Specifikationer
Kemiskt namn eller material Equilenin
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C18H18O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,16,19H,6-9H2,1H3/t16-,18-/m0/s1
Formel vikt 266.1307
Procent renhet >95%