missing translation for 'onlineSavingsMsg'
Läs mer

Equilin 3-O-beta-D-Glucuronide Sodium Salt, TRC

Produktkod. 30378475
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378475 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30378475 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE59281010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Equilin 3-O-beta-D-Glucuronide Sodium Salt
CAS 27610-12-4
Molekylformel C24H27NaO8
Kvantitet 10 mg
Synonym 17-Oxoestra-1,3,5(10),7-tetraen-3-yl β-D-Glucopyranosiduronic Acid Monosodium Salt
InChI formel InChI=1S/C24H28O8.Na/c1-24-9-8-14-13-5-3-12(10-11(13)2-4-15(14)16(24)6-7-17(24)25)31-23-20(28)18(26)19(27)21(32-23)22(29)30;/h3-5,10,14,16,18-21,23,26-28H,2,6-9H2,1H3,(H,29,30);/q;+1/p-1/t14-,16+,18+,19+,20-,21+,23-,24+;/m1./s1
LEDER [Na+].C[C@]12CC[C@H]3C(=CCc4cc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-])ccc34)[C@@H]1CCC2=O
Molekylvikt (g/mol) 466.46
Formel vikt 466.1604
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.