Learn More
Thermo Scientific™ Equine Genotypes Panel 1.1 Kit
Equine Genotypes Panel 1.1 Kit
Brand:  Thermo Scientific™ F850S
Additional Details : Weight : 0.04210kg
 View more versions of this product
Product Code. 15504404
  • 10790.00 SEK / 100 reactions
EU Stock 33
Add to basket
Description
Description
Thermo Scientific Equine Genotypes Panel 1.1 is a reagent kit for horse parentage testing and identification using short tandem repeats (STRs).
The Equine Genotypes Panel 1.1 provides all the necessary reagents for amplification of the 17 microsatellite loci. The kit contains master mix and primer mix allows co-amplification of the microsatellites in a single multiplex PCR reaction. Equine control DNA is provided to verify acceptable PCR and electrophoresis conditions. Reagents and protocols of the Equine Genotypes Panel 1.1. kit have been optimized to deliver similar peak sizes (amplification yields) for each loci. Allele callings obtained with this kit represent the true alleles of an individual, instead of 'plus-A peaks' or 'split peaks' typically interpreted when using e.g. a conventional Taq DNA polymerase. The kit employs Phusion Hot Start DNA Polymerase that has a proofreading activity and eliminates the 'plus-A peaks'. One primer from each primer pair is end-labelled with a fluorescent dye. After PCR, the fragments are separated and detected in a single electrophoresis injection, using an automated capillary electrophoresis instrument.
The Equine Genotypes Panel 1.1 allows co-amplification of the 17 microsatellites in a single multiplex PCR reaction. The Panel encompasses the following loci: VHL20, HTG4, AHT4, HMS7, HTG6, AHT5, HMS6, ASB23, ASB2, HTG10, HTG7, HMS3, HMS2, ASB17, LEX3, HMS1 and CA425. These include the nine loci recommended by the Equine Genetics and Thoroughbred Parentage Testing Standardization Committee of the International Society for Animal Genetics (ISAG) and eight additional loci commonly used for horse parentage testing and identification.
Highlights
  • Single multiplex PCR of 17 STR loci
  • Master mix and Primer mix provided for easy pipetting
  • Control DNA provided for verification of PCR and electrophoresis conditions
  • Allele calls represents true alleles (no plus-A peaks)
  • Specifications
    Specifications
    Equine Genotypes Panel 1.1 Kit, 100 rxns, Capillary Electrophoresis Detection Method, Bovine Genotyping and Parentage Testing, STR (Short Tandem Repeat) Analysis, Store at –20°C, Used For Animal Health, PCR Applications
    Animal health, PCR and real-time PCR, PCR genotyping, industrial and applied science
    Equine Genotypes Panel 1.1 Kit
    100 rxn(s)
    Capillary electrophoresis
    1.1mL Equine genotypes master mix, 1.1mL equine genotypes panel 1.1 primer mix, 30μL equine genotypes control DNA001
    100 rxn(s)
    –20°C
    Videos
    For Research Use Only. Not for use in diagnostic procedures.