Läs mer
Tocris Bioscience™ ER 50891
GREENER_CHOICE
Produktkod. 18074332
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18074332 10 mg
18054312 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18074332 Leverantör Tocris Bioscience™ Leverantörsnummer 3823/10
Selective RARα antagonist
Antagonist of RARα receptors (IC50 = 31.2 nM). Displays selective affinity for RARα (relative IC50 values are 1.8, 432 and 535 nM for RARα, RARγ and RARβ respectively).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 187400-85-7
Molekylformel C29H24N2O2
Specifikationer
Kemiskt namn eller material ER 50891
Kvantitet 10 mg
Formel vikt 432.51
Renhet >99%
Molekylvikt (g/mol) 432.523