Learn More
ERAP2 Mouse anti-Human, Unconjugated, Clone: 2B11A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67477-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The ERAP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ERAP2. This antibody recognizes human, rat antigen. The ERAP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ERAP2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Western Blot, ELISA | |
2B11A2 | |
Unconjugated | |
20 μL | |
64167 | |
IgG2b |