missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC6L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ERCC6L |
---|---|
Concentration | 0.43 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16834974
|
Proteintech
15688-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16824974
|
Proteintech
15688-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ERCC6L is a DNA helicase that acts as an essential component of the spindle assembly checkpoint. ERCC6L contributes to the mitotic checkpoint by recruiting MAD2 to kinetochores and monitoring tension on centromeric chromatin. ERCC6L acts as a tension sensor that associates with catenated DNA which is stretched under tension until it is resolved during anaphase.Specifications
ERCC6L | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q2NKX8, Q8BHK9 | |
236930, 317252, 54821 | |
ERCC6L Fusion Protein Ag8223 | |
Primary | |
-20°C | |
ERCC6L |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ERCC6L, PICH, Tumor antigen BJ HCC 15 | |
Ercc6l | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |