missing translation for 'onlineSavingsMsg'
Learn More

ERCC6L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15688-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824974

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ERCC6L is a DNA helicase that acts as an essential component of the spindle assembly checkpoint. ERCC6L contributes to the mitotic checkpoint by recruiting MAD2 to kinetochores and monitoring tension on centromeric chromatin. ERCC6L acts as a tension sensor that associates with catenated DNA which is stretched under tension until it is resolved during anaphase.
Specifications

Specifications

ERCC6L
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q2NKX8, Q8BHK9
ERCC6L, PICH, Tumor antigen BJ HCC 15
Rabbit
IgG
150 μL
-20°C
Primary
236930, 317252, 54821
Western Blot
Unconjugated
ERCC6L
Liquid
Ercc6l
ERCC6L Fusion Protein Ag8223
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers