Läs mer
Erdosteine Methyl Ester, Mikromol™
Produktkod. 17963006
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17963006 25 mg
17904546 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17963006 Leverantör Mikromol Leverantörsnummer MM0544.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Erdosteine Methyl Ester
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Erdosteine Methyl Ester
CAS 1667709-77-4
Analyt- eller komponentnamn Erdosteine Methyl Ester
Frakt skick Room Temperature
Molekylformel C9 H13 N O4 S2
Certifikat och efterlevnad ISO 9001
Synonym Erdosteine Methyl Ester, 2-[[2-Oxo-2-[(tetrahydro-2-oxo-3-thienyl)amino]ethyl]thio]-acetic acid methyl ester
InChI formel InChI=1S/C9H13NO4S2/c1-14-8(12)5-15-4-7(11)10-6-2-3-16-9(6)13/h6H,2-5H2,1H3,(H,10,11)
LEDER COC(=O)CSCC(=O)NC1CCSC1=O
IUPAC-namn methyl 2-[2-oxo-2-[(2-oxothiolan-3-yl)amino]ethyl]sulfanylacetate
Molekylvikt (g/mol) 263.33
Kvantitet 25 mg
Formel vikt 263.0286 g/mol