Läs mer
Erdosteine Methyl Ester, TRC
Produktkod. 30374540
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30374540 250 mg
1 options
Produktkod. 30374540 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE596080250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Erdosteine Methyl Ester
CAS 1667709-77-4
Molekylformel C9 H13 N O4 S2
Kvantitet 250 mg
Synonym 2-[[2-Oxo-2-[(tetrahydro-2-oxo-3-thienyl)amino]ethyl]thio]-acetic acid methyl ester
InChI formel InChI=1S/C9H13NO4S2/c1-14-8(12)5-15-4-7(11)10-6-2-3-16-9(6)13/h6H,2-5H2,1H3,(H,10,11)
LEDER COC(=O)CSCC(=O)NC1CCSC1=O
IUPAC-namn methyl 2-[2-oxo-2-[(2-oxothiolan-3-yl)amino]ethyl]sulfanylacetate
Molekylvikt (g/mol) 263.33
Formel vikt 263.0286