Läs mer
Erdosteine, TRC
Produktkod. 30012059
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012059 1 g
30011797 5 g
30006566 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012059 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5960501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84611-23-4
Molekylformel C8 H11 N O4 S2
Molekylvikt (g/mol) 249.31
Synonym Erdosteine;[[2-Oxo-2-[(tetrahydro-2-oxo-3-thienyl)amino]ethyl]thio]acetic acid, Dithiosteine, Erdostin, RV 144, Secresolv
IUPAC-namn 2-[2-oxo-2-[(2-oxothiolan-3-yl)amino]ethyl]sulfanylacetic acid
LEDER OC(=O)CSCC(=O)NC1CCSC1=O
Specifikationer
Kemiskt namn eller material Erdosteine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C8H11NO4S2/c10-6(3-14-4-7(11)12)9-5-1-2-15-8(5)13/h5H,1-4H2,(H,9,10)(H,11,12)
Formel vikt 249.0129
Procent renhet >95%