missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T, Pure, C.I.14645, Metal Indicator, Fisher Chemical™
Click to view available options
Quantity:
25 g
100 g
Description
Eriochrome Black T, Pure, C.I.14645, Metal Indicator
Specifications
IUPAC Name | (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium |
Formula Weight | 461.38g/mol |
Packaging | Glass Jar |
Color | Black |
pH | 3.7 |
Quantity | 25 g |
Physical Form | Powder |
Safety and Handling
Product Identifier
- Eriochrome Black T
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'