missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ERK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577818
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
A 1:1,000 dilution of PA5-77818 was sufficient for detection of ERK1/2 in 20 μg of HeLa cell lysate by ECL immunoblot analysis. Detects approximately 44kda (ERK1) and approximately 42kDa (ERK2).
MAPK3 (ERK1) is a serine/threonine kinase that is an important activator of p90, RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway. ERK1 is widely expressed and involved in the regulation of meiosis, mitosis, and post-mitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. Functionally, ERK1 is involved with a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. ERK1 is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms of ERK1 have been described.
Specifikationer
| ERK1 | |
| Polyclonal | |
| Unconjugated | |
| MAPK3 | |
| ERK; Erk1; ERK-1; Ert2; Esrk1; Extracellular signal-regulated kinase 1; extracellular signal-related kinase 1; HS44KDAP; HUMKER1A; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 3; MAP kinase isoform p44; MAP kinase2; MAPK 1; MAPK 3; MAPK1; MAPK2; MAPK3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 3; mitogen-activated 3; mitogen-activated protein kinase 1; mitogen-activated protein kinase 3; MNK1; Mtap2k; p44; p44 MAP kinase; P44ERK1; p44-ERK1; P44MAPK; p44-MAPK; pp42/MAP kinase; PRKM3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 101123008, 26417, 50689, 531391, 5595 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| P21708, P27361, Q63844 | |
| MAPK3 | |
| A 35 residue synthetic peptide, corresponding to rat Erk1 MAP kinase with the CGG spacer group added and the peptide coupled to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Sheep, Chicken, Drosophila, Frog | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering