Läs mer
Erlotinib-13C6 Hydrochloride, TRC
Produktkod. 30370598
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30370598 1 mg
1 options
Produktkod. 30370598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE6250031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Erlotinib-13C6 Hydrochloride
CAS 1210610-07-3
Molekylformel C1613C6H23N3O4 • HCl
Kvantitet 1 mg
Synonym N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine-13C6 Hydrochloride, 6,7-Bis(2-methoxyethoxy)-4-(3-ethynylanilino)quinazoline-13C6 Hydrochloride, [6,7-Bis(2-methoxyethoxy)quinazolin-4-yl]-(3-ethynylphenyl)amine-13C6 HydrochloridCP 358774-13C6, OSI 774-13C6, Tarceva-13C6
InChI formel InChI=1S/C22H23N3O4.ClH/c1-4-16-6-5-7-17(12-16)25-22-18-13-20(28-10-8-26-2)21(29-11-9-27-3)14-19(18)23-15-24-22;/h1,5-7,12-15H,8-11H2,2-3H3,(H,23,24,25);1H/i5+1,6+1,7+1,12+1,16+1,17+1;
LEDER C#C[13C]1=[13CH][13CH]=[13CH][13C](NC2=NC=NC3=CC(OCCOC)=C(OCCOC)C=C32)=[13CH]1.Cl
IUPAC-namn N-(3-ethynylphenyl-1,2,3,4,5,6-13C6)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride
Molekylvikt (g/mol) 435.85
Formel vikt 435.17