Läs mer
Erlotinib Hydrochloride (Tarceva), TRC
Produktkod. 18073930
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18073930 100 mg
18060340 50 mg
18003550 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18073930 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE625000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 183319-69-9
Molekylformel C22 H23 N3 O4 . Cl H
Molekylvikt (g/mol) 429.9
Synonym Erlotinib hydrochloride, 4-Quinazolinamine, N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)-, hydrochloride (1:1), 4-Quinazolinamine, N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)-, monohydrochloride (9CI), 6,7-Bis(2-methoxyethoxy)-4-(3-ethynylanilino)quinazoline hydrochloride, CP 358774, OSI 774, Tarceva, [6,7-Bis(2-methoxyethoxy)quinazolin-4-yl]-(3-ethynylphenyl)amine hydrochloride
IUPAC-namn N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine;hydrochloride
LEDER Cl.COCCOc1cc2ncnc(Nc3cccc(c3)C#C)c2cc1OCCOC
Specifikationer
Kemiskt namn eller material Erlotinib, Hydrochloride Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C22H23N3O4.ClH/c1-4-16-6-5-7-17(12-16)25-22-18-13-20(28-10-8-26-2)21(29-11-9-27-3)14-19(18)23-15-24-22;/h1,5-7,12-15H,8-11H2,2-3H3,(H,23,24,25);1H
Formel vikt 429.1455
Procent renhet >95%