missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Human ERN1, GST Tag Recombinant Protein

Produktkod. p-7181076
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
Förpackningsstorlek:
100µg
10µg
1mg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15761628

Brand: Invitrogen™ A30971

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This ERN1 recombinant human protein (468-end) was expressed in insect cells. ERN1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for ERN1 is NP_001424.3.

ERN1 serine/threonine kinase or endoplasmic reticulum to nucleus signaling 1 is the ER to nucleus signaling 1 protein, a human homologue of the yeast Ire1 gene product which possesses intrinsic kinase activity and an endoribonuclease activity and it is important in endoplasmic reticulum-based stress signals. ERN1 controls IRE1 proteolysis in mammalian cells. ERN1 has a critical function in extraembryonic cells that is essential for fetal viability. Specifically, IRE1 acts as the sensor of unfolded proteins in the ER that initiates transmittance of the unfolded protein signal from the ER to the nucleus by splicing XBP1 mRNA converting it into a potent unfolded-protein response transcriptional activator. Null mutants are viable, but IRE1 is essential for viability under stress conditions that cause unfolded proteins to accumulate in the ER. Depletion of IRE1p through the expression of a dominant negative form of IRE1p has no effect on transfected cells, but cell death via apoptosis occurs under stress conditions that cause unfolded proteins to accumulate in the ER.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer O75460
Koncentration See Label
För användning med (applikation) Kinase Assay
Formulering 50mM tris HCl with 25% glycerol, 10mM glutathione, 150mM NaCl, 0.1mM PMSF, 0.25mM DTT, 0.1mM EDTA and no preservative; pH 7.5
Gen-ID (Entrez) 2081
Molekylvikt (g/mol) 84.1 kDa
Namn Human ERN1, GST Tag
pH-intervall 7.5
Reningsmetod Purified
Kvantitet 10 μg
Förvaringskrav -80°C, Avoid Freeze/Thaw Cycles
Regulatorisk status RUO
Gene Alias 9030414B18Rik; AI225830; C85377; endoplasmic reticulum (ER) to nucleus signalling 1; endoplasmic reticulum to nucleus signaling 1; endoplasmic reticulum to nucleus signalling 1; endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; FLJ30999; hIRE1p; inositol-requiring 1; inositol-requiring 1 alpha; inositol-requiring enzyme 1; inositol-requiring protein 1; IRE1; IRE1a; IRE1a antibody; Ire1alpha; Ire1-alpha; Ire1p; IRE1p antibody; MGC163277; MGC163279; protein kinase/endoribonuclease; RGD1559716; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1; serine/threonine-protein kinase/endoribonuclease IRE1-like
Vanligt namn IRE1 alpha
Gensymbol ERN1
KinaseFamily IRE Family
Protein Längd 468-end
Produkttyp Protein
Konjugera Unconjugated
Art Human
Rekombinant Recombinant
Protein Tag GST-tag
Sekvens 468-end
Innehåll och lagring -80°C, Avoid Freeze/Thaw Cycles
Frakt skick Dry Ice
Uttryckssystem Insect cells
Proteinfamilj Kinases & Inhibitors
Proteinform Recombinant, Active, Catalytic
Form Liquid
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.