missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin, ∽98%, MP Biomedicals™

Catalog Number 11441631
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
114-07-8
C37H67NO13
733.94
MFCD00084654
ULGZDMOVFRHVEP-RWJQBGPGSA-N
12560
CHEBI:42355
(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
This item is not returnable. View return policy
114-07-8
C37H67NO13
733.94
MFCD00084654
ULGZDMOVFRHVEP-RWJQBGPGSA-N
12560
CHEBI:42355
(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

Erythromycin ∽98%, White Powder

  • It Used in cell culture applications at approximately 100 mg/liter.
  • Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth.
  • Erythromycin resistance can be induced in bacteria.
  • Erythromycin is a macrolide antibiotic that bind to the 50S ribosomal subunit and inhibit protein synthesis.
  • This is an antimicrobial substance produced by the growth of certain strains of Streptomyces erythreus.
  • It is a mixture consisting largely of erythromycin A with lesser amounts of erythromycins B and C.
Type Erythromycin
Physical Form Powder
Assay Percent Range ∽ 98%
Synonym E-mycin, Erycin
Formula Weight 733.95
Color White
Quantity 25 g
Chemical Name or Material Erythromycin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'