Läs mer
Erythromycin Ethylsuccinate, MedChemExpress
Produktkod. 16471446
Encompass_Preferred
Change view
Kvantitet:
200 mg
Produktkod. Kvantitet
16471446 200 mg
1 options
Produktkod. 16471446 Leverantör Medchem Express Leverantörsnummer HYB0957
MedChemExpress Erythromycin Ethylsuccinate is an antibiotic useful for the treatment of a number of bacterial infections, has an antimicrobial spectrum similar to or slightly wider than that of penicillin. Erythromycin Ethylsuccinate has antiviral activity against HIV-1.
Erythromycin Ethylsuccinate is an antibiotic useful for the treatment of a number of bacterial infections, has an antimicrobial spectrum similar to or slightly wider than that of penicillin. Erythromycin Ethylsuccinate has antiviral activity against HIV-1.
Specifikationer
Kemiskt namn eller material Erythromycin Ethylsuccinate
Fysisk form Powder
CAS 1264-62-6
Färg White
Kvantitet 200 mg
Anteckningar om renhetsgrad Research
Molekylformel C43H75NO16
För användning med (applikation) COVID-19-anti-virus
Synonym Erythromycin ethyl succinate EES
Löslighetsinformation DMSO : ≥ 50 mg/mL (58.00 mM) ∣Ethanol : ≥ 33.33 mg/mL (38.66 mM)
LEDER O=C(CCC(OCC)=O)O[C@H]([C@H]1N(C)C)[C@](O[C@H](C)C1)([H])O[C@@H]([C@](O)(C[C@H](C([C@@H]([C@@H](O)[C@@]2(O)C)C)=O)C)C)[C@H]([C@@H]([C@H](C(O[C@@H]2CC)=O)C)O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)C
Molekylvikt (g/mol) 862.05
Formel vikt 862.05